Anti-GJC1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB135474-100EA 550 EUR
ABCAAB135474-100
Anti-GJC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GJC1 (gap junction protein, gamma 1, 45kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR