Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: FN50]

Supplier: Abcam
ABCAAB135930-50EA 0 EUR
ABCAAB135930-50
Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: FN50]
Antibodies
Type: Primary
Antigen: CD69 (CD69 molecule)
Clonality: monoclonal
Clone: FN50
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR