Anti-P2RY14 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB136264-100EA 510 EUR
ABCAAB136264-100
Anti-P2RY14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: P2RY14 (purinergic receptor P2Y, G-protein coupled, 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR