Anti-SERCA2 ATPase Rabbit Monoclonal Antibody

Supplier: Abcam
ABCAAB137020-100EA 580 EUR
ABCAAB137020-100
Anti-SERCA2 ATPase Rabbit Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SERCA2 ATPase
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR