Anti-SLC9A6 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137185-100EA 530 EUR
ABCAAB137185-100
Anti-SLC9A6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC9A6 (solute carrier family 9, subfamily A (NHE6, cation proton antiporter 6), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR