Anti-RPRD1B Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137246-100EA 510 EUR
ABCAAB137246-100
Anti-RPRD1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPRD1B (regulation of nuclear pre-mRNA domain containing 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR