Anti-LYN Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137338-50EA 560 EUR
ABCAAB137338-50
Anti-LYN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR