Anti-CXCR6 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137344-50EA 550 EUR
ABCAAB137344-50
Anti-CXCR6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR