Anti-Sarcomeric Alpha Actinin Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137346-50EA 600 EUR
ABCAAB137346-50
Anti-Sarcomeric Alpha Actinin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Sarcomeric Alpha Actinin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR