Anti-CAMK1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137409-50EA 0 EUR
ABCAAB137409-50
Anti-CAMK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CAMK1 (calcium/calmodulin-dependent protein kinase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR