Anti-KISS1R Rabbit Polyclonal Antibody
Supplier: Abcam
ABCAAB137483-100EA 550 EUR
ABCAAB137483-100
Anti-KISS1R Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KISS1R (KISS1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human