Anti-CTSF Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137516-50EA 0 EUR
ABCAAB137516-50
Anti-CTSF Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CTSF (cathepsin F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR