Anti-TRIB3 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137526-50EA 560 EUR
ABCAAB137526-50
Anti-TRIB3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIB3 (tribbles pseudokinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR