Anti-GPT Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137531-50EA 550 EUR
ABCAAB137531-50
Anti-GPT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPT (glutamic-pyruvate transaminase (alanine aminotransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR