Anti-CYP1B1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137562-100EA 550 EUR
ABCAAB137562-100
Anti-CYP1B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP1B1 (cytochrome P450, family 1, subfamily B, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR