Anti-Nuclear Factor Erythroid 2 Related Factor 1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137572-50EA 530 EUR
ABCAAB137572-50
Anti-Nuclear Factor Erythroid 2 Related Factor 1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Nuclear Factor Erythroid 2 Related Factor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR