Anti-T Plastin Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137585-50EA 585 EUR
ABCAAB137585-50
Anti-T Plastin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: T Plastin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR