Anti-RASGRP2 Rabbit Polyclonal Antibody
Supplier: Abcam
ABCAAB137608-50EA 550 EUR
ABCAAB137608-50
Anti-RASGRP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RASGRP2 (RAS guanyl releasing protein 2 (calcium and DAG-regulated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse