Anti-SLC25A22 Rabbit Polyclonal Antibody
Supplier: Abcam
ABCAAB137614-100EA 510 EUR
ABCAAB137614-100
Anti-SLC25A22 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC25A22 (solute carrier family 25 (mitochondrial carrier: glutamate), member 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse