Anti-FSHR Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137695-100EA 560 EUR
ABCAAB137695-100
Anti-FSHR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FSHR (follicle stimulating hormone receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR