Anti-Fibronectin Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137720-100EA 550 EUR
ABCAAB137720-100
Anti-Fibronectin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR