Anti-TLR3 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137722-100EA 575 EUR
ABCAAB137722-100
Anti-TLR3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR