Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137749-100EA 575 EUR
ABCAAB137749-100
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HMOX1 (heme oxygenase (decycling) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR