Anti-PPP2R1A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137825-100EA 560 EUR
ABCAAB137825-100
Anti-PPP2R1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR