Anti-Fibrinopeptide B Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137830-100EA 550 EUR
ABCAAB137830-100
Anti-Fibrinopeptide B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Fibrinopeptide B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR