Anti-SCN4A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB138079-100EA 530 EUR
ABCAAB138079-100
Anti-SCN4A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SCN4A (sodium channel, voltage-gated, type IV, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR