Anti-IRF3 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB138449-100EA 550 EUR
ABCAAB138449-100
Anti-IRF3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR