Anti-LMNB2 Rabbit Monoclonal Antibody

Supplier: Abcam
ABCAAB138516-100EA 545 EUR
ABCAAB138516-100
Anti-LMNB2 Rabbit Monoclonal Antibody
Antibodies
Type: Primary
Antigen: LMNB2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR