Anti-LCK Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB138524-100EA 470 EUR
ABCAAB138524-100
Anti-LCK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LCK (lymphocyte-specific protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR