Anti-Phospholipase A2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB139388-50EA 560 EUR
ABCAAB139388-50
Anti-Phospholipase A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Phospholipase A2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR