Anti-KEAP1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB139729-100EA 495 EUR
ABCAAB139729-100
Anti-KEAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KEAP1 (kelch-like ECH-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR