Anti-SGSM2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB139811-100EA 510 EUR
ABCAAB139811-100
Anti-SGSM2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SGSM2 (small G protein signaling modulator 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR