Anti-PSMD14 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB140496-100EA 550 EUR
ABCAAB140496-100
Anti-PSMD14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSMD14 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR