Anti-PSMC4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB140515-100EA 550 EUR
ABCAAB140515-100
Anti-PSMC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSMC4 (proteasome (prosome, macropain) 26S subunit, ATPase, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR