Anti-ALPHA 1,2 MAN Rabbit Monoclonal Antibody

Supplier: Abcam
ABCAAB140613-100EA 565 EUR
ABCAAB140613-100
Anti-ALPHA 1,2 MAN Rabbit Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ALPHA 1,2 MAN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR