Anti-ACKR4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB140784-50EA 550 EUR
ABCAAB140784-50
Anti-ACKR4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACKR4 (atypical chemokine receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR