Anti-LOXL2 Rabbit Polyclonal Antibody
Supplier: Abcam
ABCAAB140833-50EA 560 EUR
ABCAAB140833-50
Anti-LOXL2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LOXL2 (lysyl oxidase-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human