Anti-PRLHR Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB140949-50EA 510 EUR
ABCAAB140949-50
Anti-PRLHR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRLHR (prolactin releasing hormone receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR