Anti-MAD2L1 Rabbit Monoclonal Antibody

Supplier: Abcam
ABCAAB150371-100EA 525 EUR
ABCAAB150371-100
Anti-MAD2L1 Rabbit Monoclonal Antibody
Type: Primary
Antigen: MAD2L1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR