Anti-Protein C Rabbit Monoclonal Antibody

Supplier: Abcam
ABCAAB150373-100EA 570 EUR
ABCAAB150373-100
Anti-Protein C Rabbit Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Protein C
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR