Anti-PTGDR2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB150632-50EA 550 EUR
ABCAAB150632-50
Anti-PTGDR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTGDR2 (prostaglandin D2 receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR