Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Supplier: Biotium
20103-1EA 137 EUR
20103-1 20103.
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR