Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)

Supplier: Biotium
20157-1EA 137 EUR
20157-1 20157.
Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR