Anti-IgG Goat Polyclonal Antibody (CF™ 405M)

Supplier: Biotium

20180-1EA 110 EUR
20180-1 20180.
Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 405M Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR