Anti-6X His tag Mouse Monoclonal Antibody (CF®640R)

Supplier: Biotium

20237.EA 518 EUR
20237.
Anti-6X His tag Mouse Monoclonal Antibody (CF®640R)
Antibodies
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR