Anti-HA tag Rabbit Polyclonal Antibody (CF®594)

Supplier: Biotium

20239.EA 518 EUR
20239.
Anti-HA tag Rabbit Polyclonal Antibody (CF®594)
Antibodies
Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR