Anti-IgG Donkey Polyclonal Antibody (CF™ 680)

Supplier: Biotium

20278-1EA 137 EUR
20278-1 20278.
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR