Anti-IgG2b Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium
20327-1EA 154 EUR
20327-1 20327.
Anti-IgG2b Goat Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR