Anti-CD4 Mouse Monoclonal Antibody [clone: CA-4]

Supplier: Boster
BSBTMA1016EA 550 EUR
BSBTMA1016
Anti-CD4 Mouse Monoclonal Antibody [clone: CA-4]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR