Anti-Cytokeratin Peptide 7 Mouse Monoclonal Antibody [clone: CK-7]

Supplier: Boster
BSBTMA1024EA 550 EUR
BSBTMA1024
Anti-Cytokeratin Peptide 7 Mouse Monoclonal Antibody [clone: CK-7]
Antibodies
Type: Primary
Antigen: Cytokeratin Peptide 7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR